Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps-14
Entrez Gene ID
176006
Gene Name
40S ribosomal protein S14
Gene Aliases
CELE_F37C12.9
Chromosome Location
Chr.III: 7178980 - 7181523 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066171.9 NP_498572.1 5 - 6 930 58

View Full Product Details