Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pfd-5
Entrez Gene ID
176013
Gene Name
Probable prefoldin subunit 5
Gene Aliases
CELE_R151.9
Chromosome Location
Chr.III: 7198151 - 7198884 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066181.6 NP_498582.1 2 - 3 220 87
GenBank mRNA AF303258.1 - 2 - 3 220 87

View Full Product Details