Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- adr-2
- Entrez Gene ID
-
176022
- Gene Name
- Double-stranded RNA-specific adenosine deaminase adr-2
- Gene Aliases
-
CELE_T20H4.4
- Chromosome Location
-
Chr.III: 7231039 - 7232882 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066193.5 |
NP_498594.1 |
3 - 4 |
981 |
68 |
GenBank mRNA |
AF051275.1 |
- |
3 - 4 |
1003 |
68 |
- Gene Symbol
- R151.10
- Entrez Gene ID
-
24104817
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_R151.10
- Chromosome Location
-
Chr.III: 7230054 - 7232884 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382910.1 |
|
3 - 4 |
983 |
68 |
View Full Product Details