Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
F08F8.11
Entrez Gene ID
27307933
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F08F8.11
Chromosome Location
Chr.III: 7365902 - 7366448 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066227.3 NP_498628.2 1 - 2 90 148

View Full Product Details