Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mml-1
Entrez Gene ID
176050
Gene Name
protein-coding
Gene Aliases
CELE_T20B12.6
Chromosome Location
Chr.III: 7370127 - 7374731 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379792.2 NP_001367742.1 4 - 5 838 69
NM_066230.6 NP_498631.2 4 - 5 844 69
GenBank mRNA AF264757.1 - 4 - 5 838 69

View Full Product Details