Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hmg-4
Entrez Gene ID
176052
Gene Name
FACT complex subunit SSRP1-A
Gene Aliases
CELE_T20B12.8
Chromosome Location
Chr.III: 7379143 - 7381609 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066232.6 NP_498633.1 3 - 4 1816 116

View Full Product Details