Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mup-4
Entrez Gene ID
176060
Gene Name
Transmembrane matrix receptor MUP-4
Gene Aliases
CELE_K07D8.1
Chromosome Location
Chr.III: 7479680 - 7489105 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066244.7 NP_498645.1 17 - 18 5456 97

View Full Product Details