Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cux-7
- Entrez Gene ID
-
176066
- Gene Name
- Clk-2 Upstream, human gene XE7 related
- Gene Aliases
-
CELE_C07H6.8
- Chromosome Location
-
Chr.III: 7518627 - 7520805 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066250.8 |
NP_498651.2 |
3 - 4 |
748 |
121 |
- Gene Symbol
- clk-2
- Entrez Gene ID
-
176065
- Gene Name
- Telomere length regulation protein clk-2
- Gene Aliases
-
CELE_C07H6.6
- Chromosome Location
-
Chr.III: 7515223 - 7520845 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382917.1 |
|
3 - 4 |
788 |
121 |
View Full Product Details