Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdh-3
Entrez Gene ID
176085
Gene Name
Cadherin-3
Gene Aliases
CELE_ZK112.7
Chromosome Location
Chr.III: 7741229 - 7754448 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066286.6 NP_498687.2 18 - 19 7717 97

View Full Product Details