Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-5
Entrez Gene ID
176093
Gene Name
protein-coding
Gene Aliases
CELE_C08C3.1
Chromosome Location
Chr.III: 7814374 - 7817141 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025995.3 NP_001021166.1 1 - 2 193 100
NM_001025996.3 NP_001021167.1 1 - 2 199 100
NM_066296.5 NP_498697.3 1 - 2 193 100
GenBank mRNA L19247.1 - 1 - 2 191 100

View Full Product Details