Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-15
Entrez Gene ID
176114
Gene Name
Nuclear pore complex protein 15
Gene Aliases
CELE_C29E4.4
Chromosome Location
Chr.III: 7929013 - 7933055 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066323.6 NP_498724.4 1 - 2 122 69

View Full Product Details