Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kle-2
Entrez Gene ID
176116
Gene Name
Condensin-2 complex subunit kle-2
Gene Aliases
CELE_C29E4.2
Chromosome Location
Chr.III: 7941717 - 7945594 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066326.8 NP_498727.2 5 - 6 2017 80

View Full Product Details