Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-36
Entrez Gene ID
176128
Gene Name
Protein lin-36
Gene Aliases
CELE_F44B9.6
Chromosome Location
Chr.III: 8016648 - 8020879 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066346.6 NP_498747.2 3 - 4 794 70
GenBank mRNA AF104917.1 - 3 - 4 797 70

View Full Product Details