Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apc-2
Entrez Gene ID
176139
Gene Name
Anaphase-promoting complex subunit 2
Gene Aliases
CELE_K06H7.6
Chromosome Location
Chr.III: 8080570 - 8083701 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066361.6 NP_498762.2 12 - 13 1714 118

View Full Product Details