Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- fbxl-1
- Entrez Gene ID
-
3564805
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C02F5.7
- Chromosome Location
-
Chr.III: 8226416 - 8232572 on Build WBcel235
- Gene Symbol
- rfs-1
- Entrez Gene ID
-
24104262
- Gene Name
- RAD51-like protein 1
- Gene Aliases
-
CELE_C30A5.2
- Chromosome Location
-
Chr.III: 8226417 - 8232559 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382934.1 |
|
2 - 3 |
632 |
134 |
View Full Product Details