Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
knl-1
Entrez Gene ID
176164
Gene Name
Kinetochore null protein 1
Gene Aliases
CELE_C02F5.1
Chromosome Location
Chr.III: 8250632 - 8253943 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066410.6 NP_498811.1 2 - 3 241 97

View Full Product Details