Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mig-10
Entrez Gene ID
176168
Gene Name
protein-coding
Gene Aliases
CELE_F10E9.6
Chromosome Location
Chr.III: 8279101 - 8305837 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379826.1 NP_001367746.1 3 - 4 449 151
NM_001382935.1 3 - 4 453 151

View Full Product Details