Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mig-10
Entrez Gene ID
176168
Gene Name
protein-coding
Gene Aliases
CELE_F10E9.6
Chromosome Location
Chr.III: 8279101 - 8305837 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379826.1 NP_001367746.1 6 - 7 867 81
NM_001382935.1 6 - 7 871 81
NM_066420.6 NP_498821.2 4 - 5 501 81

View Full Product Details