Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lap-1
Entrez Gene ID
176185
Gene Name
Leucine aminopeptidase 1
Gene Aliases
CELE_ZK353.6
Chromosome Location
Chr.III: 8399168 - 8401928 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066453.4 NP_498854.1 4 - 5 962 95

View Full Product Details