Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ubxn-4
Entrez Gene ID
176187
Gene Name
UBX domain-containing protein 4
Gene Aliases
CELE_ZK353.8
Chromosome Location
Chr.III: 8403718 - 8405431 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382940.1 3 - 4 854 101

View Full Product Details