Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ufbp-1
Entrez Gene ID
176194
Gene Name
protein-coding
Gene Aliases
CELE_ZK1236.7
Chromosome Location
Chr.III: 8439531 - 8443607 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268030.3 NP_001254959.1 2 - 3 243 63
NM_001379832.1 NP_001368103.1 2 - 3 174 63

View Full Product Details