Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ufbp-1
Entrez Gene ID
176194
Gene Name
protein-coding
Gene Aliases
CELE_ZK1236.7
Chromosome Location
Chr.III: 8439531 - 8443607 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268030.3 NP_001254959.1 4 - 5 873 101
NM_001379832.1 NP_001368103.1 4 - 5 804 101

View Full Product Details