Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glr-1
Entrez Gene ID
176204
Gene Name
Glutamate receptor 1
Gene Aliases
CELE_C06E1.4
Chromosome Location
Chr.III: 8584163 - 8588642 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066486.8 NP_498887.2 12 - 13 2724 113
GenBank mRNA U34661.1 - 12 - 13 2803 113

View Full Product Details