Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpy-19
Entrez Gene ID
191628
Gene Name
C-mannosyltransferase dpy-19
Gene Aliases
CELE_F22B7.10
Chromosome Location
Chr.III: 8659872 - 8667482 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066508.2 NP_498909.1 9 - 10 1565 140

View Full Product Details