Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fli-1
Entrez Gene ID
176215
Gene Name
Protein flightless-1 homolog
Gene Aliases
CELE_B0523.5
Chromosome Location
Chr.III: 8675297 - 8683736 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066512.6 NP_498913.2 8 - 9 2073 127
GenBank mRNA U01183.1 - 9 - 10 2106 127

View Full Product Details