Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugtp-1
Entrez Gene ID
176227
Gene Name
UDP-galactose translocator 1
Gene Aliases
CELE_ZK370.7
Chromosome Location
Chr.III: 8746217 - 8750706 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066529.7 NP_498930.1 6 - 7 873 104

View Full Product Details