Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sma-2
Entrez Gene ID
176229
Gene Name
protein-coding
Gene Aliases
CELE_ZK370.2
Chromosome Location
Chr.III: 8749680 - 8757089 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372636.2 NP_001360602.1 3 - 4 564 84
NM_001382944.1 8 - 9 1261 84
GenBank mRNA U10327.1 - 7 - 8 1070 84
U34778.1 - 6 - 7 1026 84

View Full Product Details