Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-9
Entrez Gene ID
176256
Gene Name
protein-coding
Gene Aliases
CELE_ZK637.7
Chromosome Location
Chr.III: 8900231 - 8904459 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027844.4 NP_001023015.1 2 - 3 200 89
NM_001027845.5 NP_001023016.1 2 - 3 191 89
GenBank mRNA AF269693.1 - 2 - 3 194 89
AF269694.1 - 2 - 3 194 89

View Full Product Details