Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trxr-2
Entrez Gene ID
176259
Gene Name
Probable glutathione reductase 2
Gene Aliases
CELE_ZK637.10
Chromosome Location
Chr.III: 8913769 - 8916463 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066570.5 NP_498971.1 4 - 5 1034 92

View Full Product Details