Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nac-2
Entrez Gene ID
187898
Gene Name
protein-coding
Gene Aliases
CELE_R107.1
Chromosome Location
Chr.III: 9038779 - 9043300 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268047.2 NP_001254976.1 9 - 10 1453 104
NM_001268048.2 NP_001254977.1 8 - 9 1200 104

View Full Product Details