The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_066605.8 | NP_499006.1 | 3 - 4 | 313 | 71 |
GenBank mRNA | X13689.1 | - | - | 275 | 71 |