Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glp-1
Entrez Gene ID
176286
Gene Name
Protein glp-1
Gene Aliases
CELE_F02A9.6
Chromosome Location
Chr.III: 9092241 - 9099698 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066613.5 NP_499014.1 8 - 9 3841 67

View Full Product Details