Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dat-1
Entrez Gene ID
176304
Gene Name
Sodium-dependent dopamine transporter
Gene Aliases
CELE_T23G5.5
Chromosome Location
Chr.III: 9242914 - 9246802 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066642.6 NP_499043.1 2 - 3 200 68
GenBank mRNA AF079899.1 - 2 - 3 208 68

View Full Product Details