Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elo-4
Entrez Gene ID
183367
Gene Name
Putative fatty acid elongation protein 4
Gene Aliases
CELE_C40H1.4
Chromosome Location
Chr.III: 9332265 - 9335328 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066655.5 NP_499056.1 4 - 5 400 122

View Full Product Details