Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpar-1
Entrez Gene ID
186223
Gene Name
Histone H3-like centromeric protein cpar-1
Gene Aliases
CELE_F54C8.2
Chromosome Location
Chr.III: 9434057 - 9435100 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066672.5 NP_499073.1 2 - 3 254 89

View Full Product Details