Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rheb-1
Entrez Gene ID
176327
Gene Name
GTP-binding protein Rheb homolog 1
Gene Aliases
CELE_F54C8.5
Chromosome Location
Chr.III: 9454163 - 9456030 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066678.4 NP_499079.1 4 - 5 373 94

View Full Product Details