Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clu-1
Entrez Gene ID
176341
Gene Name
Clustered mitochondria protein homolog
Gene Aliases
CELE_F55H2.6
Chromosome Location
Chr.III: 9516629 - 9521460 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066696.8 NP_499097.1 5 - 6 3103 75

View Full Product Details