Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ced-7
Entrez Gene ID
176352
Gene Name
protein-coding
Gene Aliases
CELE_C48B4.4
Chromosome Location
Chr.III: 9570390 - 9580089 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026054.4 NP_001021225.1 2 - 3 280 93
NM_001268083.3 NP_001255012.1 2 - 3 339 93
NM_001379863.1 NP_001366984.1 1 - 2 138 93
NM_066714.4 NP_499115.2 2 - 3 288 93
GenBank mRNA AF049142.1 - 2 - 3 301 93

View Full Product Details