The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026054.4 | NP_001021225.1 | 5 - 6 | 1565 | 106 |
NM_001268083.3 | NP_001255012.1 | 5 - 6 | 1624 | 106 | |
NM_001379863.1 | NP_001366984.1 | 4 - 5 | 1423 | 106 | |
NM_066714.4 | NP_499115.2 | 5 - 6 | 1573 | 106 | |
GenBank mRNA | AF049142.1 | - | 5 - 6 | 1586 | 106 |