Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hlh-11
Entrez Gene ID
176360
Gene Name
protein-coding
Gene Aliases
CELE_F58A4.7
Chromosome Location
Chr.III: 9619711 - 9626529 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379865.1 NP_001367058.1 7 - 8 1386 58
NM_066728.7 NP_499129.2 7 - 8 1415 58
NM_066729.9 NP_499130.2 7 - 8 1408 58

View Full Product Details