Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-13
Entrez Gene ID
176364
Gene Name
Gex-3-interacting protein 13
Gene Aliases
CELE_F58A4.11
Chromosome Location
Chr.III: 9631994 - 9637116 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066733.5 NP_499134.2 3 - 4 744 129

View Full Product Details