Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iff-1
Entrez Gene ID
176373
Gene Name
protein-coding
Gene Aliases
CELE_T05G5.10
Chromosome Location
Chr.III: 9745862 - 9747274 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268091.3 NP_001255020.1 4 - 5 636 63
NM_001268092.2 NP_001255021.1 3 - 4 414 63
NM_001268093.3 NP_001255022.1 1 - 2 128 63

View Full Product Details