Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sqv-3
Entrez Gene ID
176382
Gene Name
Xylosylprotein 4-beta-galactosyltransferase
Gene Aliases
CELE_R10E11.4
Chromosome Location
Chr.III: 9778782 - 9780354 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066763.7 NP_499164.1 4 - 5 632 113
GenBank mRNA AJ005867.1 - 4 - 5 635 113

View Full Product Details