Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnx-1
Entrez Gene ID
176390
Gene Name
protein-coding
Gene Aliases
CELE_ZK632.6
Chromosome Location
Chr.III: 9814268 - 9817137 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066775.6 NP_499176.1 2 - 3 420 101

View Full Product Details