Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alg-4
Entrez Gene ID
176400
Gene Name
protein-coding
Gene Aliases
CELE_ZK757.3
Chromosome Location
Chr.III: 9887210 - 9891458 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066792.4 NP_499193.1 2 - 3 97 89

View Full Product Details