Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-3
Entrez Gene ID
186957
Gene Name
Transthyretin-like protein 3
Gene Aliases
CELE_K03H1.3
Chromosome Location
Chr.III: 9925530 - 9926750 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066801.4 NP_499202.1 2 - 3 182 114

View Full Product Details