Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alx-1
Entrez Gene ID
176410
Gene Name
protein-coding
Gene Aliases
CELE_R10E12.1
Chromosome Location
Chr.III: 9957351 - 9963171 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027542.4 NP_001022713.1 4 - 5 1200 67
NM_001027543.4 NP_001022714.1 4 - 5 1206 67
NM_066812.7 NP_499213.1 4 - 5 1206 67
GenBank mRNA U73679.1 - 4 - 5 1211 67

View Full Product Details