Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-8
Entrez Gene ID
176411
Gene Name
Prion-like-(Q/N-rich) domain-bearing protein 8
Gene Aliases
CELE_C05B5.3
Chromosome Location
Chr.III: 9996197 - 9997990 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322760.3 NP_001309437.1 3 - 4 1066 70

View Full Product Details