Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rgs-1
Entrez Gene ID
176415
Gene Name
protein-coding
Gene Aliases
CELE_C05B5.7
Chromosome Location
Chr.III: 10014188 - 10016558 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382973.1 2 - 3 620 163
GenBank mRNA AF220159.1 - 2 - 3 641 163

View Full Product Details