Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
xbx-3
Entrez Gene ID
187445
Gene Name
X-BoX promoter element regulated
Gene Aliases
CELE_M04D8.6
Chromosome Location
Chr.III: 10035945 - 10036956 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066826.2 NP_499227.1 3 - 4 570 83

View Full Product Details